missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
NOTRAX™ Marble Tuff™ Anti-Fatigue Mat
Marble Tuff™ Anti-Fatigue Floor Mat provides comfort and ergonomic support
Supplier: NOTRAX™ 511S0035GY
Description
- Durable smooth marble-pattern top surface of heavy-duty vinyl
- Easy to sweep clean or damp mop
- Dense closed cell PVC foam base provides added cushioning comfort
- Smooth attractive surface east to sweep clean
- Beveled edges on all four sides
- Thickness: 0.5 in.
Specifications
3 ft. | |
5 ft. | |
0.5 in. |
Gray | |
1 Ea. | |
Marble pattern top |