Brady™ Marking Tape Roll: High Performance Laminated Vinyl, Diagonal Stripes, Green on White
High Performance Laminated Vinyl (B-950) tape is perfect for marking obstructions, low hanging objects, pillars, passageways, aisles and other hazardous areas or objects.
Supplier: Brady™ 90020
Description
- High gloss, durable vinyl with pressure-sensitive rubber-based adhesive withstands most common chemicals, solvents, oils and water
- Encapsulated graphics are sealed beneath a protective layer of abrasion-resistant vinyl
- Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
- Designed to withstand tough indoor conditions
Specifications
Green on White |