missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Masterflex™ Compact Tubing Pumps
Masterflex™ Compact Tubing Pumps offer enhanced and versatile performance in an ultra-compact, low-maintenance design
Supplier: Masterflex™ MFLX7712230
Description
Specifications
Analog | |
1.2 to 10 rpm | |
Power cord | |
115/230 VAC |
0.002 to 1.65 mL/min | |
Yes | |
ABS plastic | |
115/230 VAC |