missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Masterflex™ Compact Tubing Pumps
Masterflex™ Compact Tubing Pumps offer enhanced and versatile performance in an ultra-compact, low-maintenance design
Supplier: Masterflex™ MFLX7712234
Description
Specifications
Analog | |
0.7 to 50 mL/min | |
Yes | |
ABS plastic | |
115/230 VAC |
No Display | |
50 to 300 rpm | |
Power cord | |
115/230 VAC |