Thermo Scientific™ Matrix™ 500μL ScrewTop Tubes
Store samples securely at low temperatures, including vapor phase Liquid Nitrogen with these ScrewTop tubes.
$1514.85 - $3073.91
Specifications
Capacity (Metric) 500 μL
Well Shape V-bottom
Product Line Matrix™
Capacity (English) 16907 fl oz.
Dnase Free Yes
View More Specs
Products 17
Catalog Number Mfr. No. Closure Color Description Packaging Fit for Purpose Price Quantity  
Catalog Number Mfr. No. Closure Color Description Packaging Fit for Purpose Price Quantity  
14754585
View Documents
Thermo Scientific™
N3743
Colorless V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready
Case for $1,755.94
14754591
View Documents
Thermo Scientific™
3743RED
Red V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready
Case for $1,812.21
14754594
View Documents
Thermo Scientific™
3743YEL
Yellow V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready
Case for $1,812.21
14754588
View Documents
Thermo Scientific™
3743BLU
Blue V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready
Case for $1,812.21
14754589
View Documents
Thermo Scientific™
3743GRE
Green V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready
Case for $1,812.21
14754592
View Documents
Thermo Scientific™
3743WHI
White V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready
Case for $1,800.15
14754590
View Documents
Thermo Scientific™
3743PUR
Purple V-Bottom ScrewTop Tubes Bulk, 480 Tubes Sterile & PCR Ready
Case for $1,812.21
14754595
View Documents
Thermo Scientific™
N3744
Colorless V-Bottom ScrewTop Tubes 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $2,563.59
14754604
View Documents
Thermo Scientific™
3744RED
Red V-Bottom ScrewTop Tubes 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $2,619.85
14754612
View Documents
Thermo Scientific™
3744YEL
Yellow V-Bottom ScrewTop Tubes 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $2,619.85
14754596
View Documents
Thermo Scientific™
3744BLU
Blue V-Bottom ScrewTop Tubes 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $2,619.85
14754599
View Documents
Thermo Scientific™
3744GRE
Green V-Bottom ScrewTop Tubes 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $2,619.85
14754606
View Documents
Thermo Scientific™
3744WHI
White V-Bottom ScrewTop Tubes 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $2,643.97
14754602
View Documents
Thermo Scientific™
3744PUR
Purple V-Bottom ScrewTop Tubes 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $2,643.97
14754614
View Documents
Thermo Scientific™
N3745
n.a. V-Bottom ScrewTop Tubes, No Caps 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $1,514.85
14754586
View Documents
Thermo Scientific™
3743AMB
Red 2D tube with red cap 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $2,748.44
14754608
View Documents
Thermo Scientific™
3744WP
Colorless V-Bottom ScrewTop Tubes with white patch 5 Latch Racks of 96 Tubes Sterile & PCR Ready
Case for $3,073.91
Description
Description
Store samples securely at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 500μL ScrewTop tubes. These tubes are molded from medical grade resins and feature a permanent, high contrast 2D barcode. Used in combination with Thermo Scientific™ VisionMate™ 2D barcode readers, manual and automated capping equipment, and labeling options, the 2D ScrewTop Tubes meet a range of manual and automated long term storage needs.
Dependable Sample Protection
  • Tubes are constructed of virgin class VI medical grade polypropylene
  • ScrewTop caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy acrylonitrile butadiene styrene (ABS) to protect tubes in case of accidental dropping
  • ScrewTop Tubes are compatible with storage down to -176°C
Secure Tracking
  • Permanent, unique 2D barcode is laser-etched on the bottom of each tube
  • 2D barcode allows you to scan and decode tubes at once without removing them from the rack, and allows data to be associated with each tube
  • Compatible with our range of VisionMate readers
Superior Storage Format
  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping / decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at a time
Stringent Quality Control
  • Every 2D barcode is scanned for readability
  • Every 2D barcode is checked against the database of all previous Thermo Scientific 2D barcodes to ensure there are zero duplicates
  • Each tube is leak tested to ensure tube integrity and sample security
  • All tubes are supplied free from DNA, RNase, DNase, PCR Inhibitors, endotoxins and cytotoxins
Specifications
Specifications
500 μL
Matrix™
Yes
V-bottom
16907 fl oz.
Sterile & PCR Ready
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'