Fisherbrand™ Maxima C Plus Gasket Sets
Replacement gasket sets for use with Maxima C vacuum pumps
Supplier: Fisherbrand™ 012573A
Description
Contain all the Viton™ O-rings, shaft seals, and gaskets needed to rebuild the pump.
†For M4C, M6C or M8C vacuum pumps designed before September 1999 (S/N before FKK090000).
Specifications
Model M2C and M4C vacuum pump | |
Viton™ O-rings, shaft seals and gaskets | |
Gasket Set |