missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ MaximaDry™ Diaphragm Vacuum Pump

Clean, reliable, oil-free vacuum pumps are ideal for low to high boiling point rotary evaporation and distillation, filtration, blotting, and solid phase extraction.

$3671.23 - $7493.67

Specifications

Maximum Pressure 15 psig
View More Specs

Products 5
Catalog Number Mfr. No. Model Flow Rate Price Quantity  
Catalog Number Mfr. No. Model Flow Rate Price Quantity  
13-880-18
View Documents
Fisherbrand™
076752029940
MaximaDry™ 20 to 6 1;15V 20 L/min.
Each for $4,825.23
 
13-880-14
View Documents
Fisherbrand™
076855043272
MaximaDry™ 20 to 75; 115V 20 L/min.
Each for $3,671.23
 
13-880-22
View Documents
Fisherbrand™
076858047708
MaximaDry™ 35 to 1.5; 115V 35 L/min.
Each for $7,493.67
 
13-880-20
View Documents
Fisherbrand™
076857029941
MaximaDry™ 35 to 6; 115V 35 L/min.
Each for $6,126.08
 
13-880-16
View Documents
Fisherbrand™
076856029940
MaximaDry™ 40 to 65; 115V 40 L/min.
Each for $5,199.13
 
Description

Description

Clean, reliable, oil-free vacuum pumps are ideal for high boiling point rotary evaporation and distillion, vacuum concentration, and larger vacuum ovens (>0.5 cu.ft.)

Quiet, reliable operation without the hazards of handling pump oil or contaminated water. Excellent replacement for rotary vane pumps and water aspirators, especially in applications where chemical vapors are encountered.

  • Withstands aggressive gases and vapors — can be used with aggressive solvents without the risk of damaging the pump
  • Single-piece PTFE coated diaphragm and molded PTFE pump heads provide chemical resistance
  • High-flow, multiport valves are made from tough FFPM fluoropolymer to withstand pumping heavy solvent vapors and liquid condensate without a loss in performance
  • Fan-cooled (TEFC) motor is rated for continuous operation and never needs lubrication
  • Compact design fits easily on a benchtop or in a fume hood
  • Setup and disassembly are quick and easy and do not require any special tools
  • Available in either 115 V/60 Hz or 230 V/50 Hz

Specifications

Specifications

15 psig
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'