Promega Maxwell™ 16 Purification Instruments DFS Item
Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
Supplier:  Promega AS1200
 View more versions of this product
Catalog No. PRAS1200
Explore available promotions
Includes: Standard Magnetic rod/plunger bar assembly, Maxwell™ 16 cartridge rack, Maxwell™ 16 Magnetic Elution rack, quick start guide
Requires : Either the SEV kit (Mfr. No. AS1200) or the LEV kit (Mfr. No. AS1250) to complete a functional instrument. Also requires Maxwell 16 Purification Kits.
Description
Description
  • Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
  • Standard Elution Volume (SEV) is 200-400μL
  • Low Elution Volume (LEV) is 30-100μL
  • Plungers and Elution Tubes for the Maxwell(R) 16 Instruments only (AS2000 and AS3000)
Certifications
All Maxwell 16 instruments are labeled GPLE (General Purpose Laboratory Equipment) in the U.S.
Specifications
Specifications
15°C to 30°C
Automated purification of genomic DNA, RNA or recombinant Proteins from a wide range of sample types
Kit
Maxwell™ 16 AS2000 instrument
Maxwell 16 instrument includes power cable and RS-232 cable for firmware upgrades. SEV Hardware Kit includes Maxwell 16 magnetic rod assembly, Maxwell 16 cartridge rack, Maxwell 16 magnetic elution rack. LEV Hardware Kit includes LEV magnetic rod assembl
1/Ea.
Product Suggestions
Product Suggestions