Promega Maxwell RSC Viral Total Nucleic Acid Purification Kit, 48 preps DFS Item
The Maxwell RSC Viral Total Nucleic Acid Purification Kit is designed for automated extraction of viral total nucleic acid (RNA and DNA) from serum, plasma or other samples using the Maxwell RSC Instrument.
Supplier:  Promega AS1330
Catalog No. PRAS1330
Explore available promotions
Description
Description
  • Automated Purification of Viral Total Nucleic Acid
  • Extracts viral total nucleic acid (RNA and DNA) from serum, plasma and other samples following brief lysis step
  • Accommodates a range of samples sizes from 100−300μl
  • Yields highly concentrated nucleic acids that are ready to use in downstream applications
Specifications
Specifications
48 Preps