Axygen™ MaxyClear Snaplock Microtubes, 1.5 mL
Ensure an efficient seal that stays closed during centrifugation. Axygen™ MaxyClear Snaplock Microtubes, 1.5mL provide excellent visibility and feature a piercing port for syringe access. Available in a wide variety of colors. Maxymum Recovery™ and sterile options available.
$52.86 - $798.30
Specifications
Max. RCF 20,000 x g
Graduations 0.1 to 1.5 mL
Cap Style Snap Cap
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Material Polypropylene
View More Specs
Products 11
Catalog Number Mfr. No. Color Sterility Packaging Price Quantity  
Catalog Number Mfr. No. Color Sterility Packaging Price Quantity  
14-222-158
Accessories View Documents
Axygen™
MCT150LC
Clear Non-sterile 250 Maxymum Recovery Tubes/Pk.
Pack of 250 Each for $54.38
Case of 10 Pack for $543.80
14-222-156
Accessories View Documents
Axygen™
MCT150CS
Clear Sterile 250 Sterile Tubes/Pk., 10 Packs/Cs.
Pack of 250 Each for $52.86
Case of 10 Pack for $528.60
14-222-154
View Documents
Axygen™
MCT150B
Blue Non-sterile 500 Tubes/Pk., 10 Packs/Cs.
Pack of 500 Each for $67.74
Case of 10 Pack for $677.40
14-222-153
Accessories View Documents
Axygen™
MCT150A
Assorted Non-sterile 500 Tubes/Pk., 10 Packs/Cs.
Pack of 500 Each for $69.41
Case of 10 Pack for $694.10
14-222-155 Edge
Accessories View Documents
Axygen™
MCT150C
Clear Non-sterile 500 Tubes/Pk., 10 Packs/Cs.
Pack of 500 Each for $60.41
Case of 10 Pack for $604.10
Edge
50-400-402
View Documents
Axygen™
MCT150G
Green Non-sterile 500 Tubes/Pk., 10 Packs/Cs.
Pack of 500 Each for $76.79
Case of 10 Pack for $767.90
50-400-406
View Documents
Axygen™
MCT150R
Red Non-sterile 500 Tubes/Pk., 10 Packs/Cs.
Pack of 500 Each for $76.79
Case of 10 Pack for $767.90
14-222-163
View Documents
Axygen™
MCT150V
Violet Non-sterile 500 Tubes/Pk., 10 Packs/Cs.
Pack of 500 Each for $66.56
Case of 10 Pack for $665.60
50-400-407
View Documents
Axygen™
MCT150SP
Spectrum Non-sterile 500 Tubes/Pk., 10 Packs/Cs.
Case for $798.30
14-222-164
View Documents
Axygen™
MCT150X
Amber Non-sterile 500 Tubes/Pk., 10 Packs/Cs.
Pack of 500 Each for $75.40
Case of 10 Pack for $754.00
50-400-410
View Documents
Axygen™
MCT150Y
Yellow Non-sterile 500 Tubes/Pk., 10 Packs/Cs.
Pack of 5000 Each for $74.28
Case of 10 Pack for $742.80
Description
Description
Ensure an efficient seal that stays closed during centrifugation. Axygen™ MaxyClear Snaplock Microtubes, 1.5mL provide excellent visibility and feature a piercing port for syringe access. Available in a wide variety of colors. Maxymum Recovery™ and sterile options available.
Axygen MaxyClear microcentrifuge tubes are available in a variety of colors. Axygen Maxymum Recovery™ surface and sterile options are also available. SnapLock microcentrifuge tubes have frosted cap surfaces and frosted side panels, ideal for labeling. Each cap features a piercing port in the center of the cap. Microcentrifuge tubes with no caps are available for some sizes.
  • 0.6 mL, 1.5 mL, and 2.0 mL clear microcentrifuge tubes are boil proof. Colored tubes are not boil-proof.
  • Co-polymer tubes offer exceptional strength and chemical resistance. Not to be autocalvable or boil-proof
  • Clear microcentrifuge tubes are boil-proof. Colored tubes are not boil-proof
  • Maximum G-force is 14,000 RCF
  • RNase-/DNase-free and nonpyrogenic
  • Sterility Assurance Level (SAL) of 10−6
  • Color - Assorted: Each pack contains a mixture of blue, green, violet, orange, red, and yellow
  • Clear microcentrifuge tubes are boil-proof. Colored tubes are not boil-proof
  • Color - Spectrum: 2 packs each of blue, green, orange, red, and yellow
Certifications
Certified RNase-, DNase- and endotoxin-safe for total sample integrity
Specifications
Specifications
20,000 x g
Snap Cap
Polypropylene
1.5 mL
0.1 to 1.5 mL
RNase, DNase, pyrogen-free
Autoclavable
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'