missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ MCDB 131 Medium, no glutamine
MCDB 131 Medium, no glutamine
Supplier: Gibco™ 10372019
Description
MCDB 131 Medium was originally developed by Knedler and Ham as reduced serum-supplemented medium for the culture of human microvascular endothelial cells (HUVEC). MCDB 131 Medium is also used with other cell types, including human omental microvascular cells, hepatocytes, myocytes, and smooth muscle cells.Using MCDB 131
MCDB 131 Medium contains many components not found in traditional basal media, such as trace elements, putrescine, adenine, thymidine, and higher levels of some amino acids and vitamins. These additions allow the medium to be supplemented with very low levels of serum or defined components. MCDB 131 Medium contains no proteins or growth factors, and is often supplemented with EGF, hydrocortisone, glutamine, and low levels of Fetal Bovine Serum (FBS). The FBS concentration must be optimized for each cell type. MCDB 131 Medium uses a sodium bicarbonate buffer system (1.176 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Dual-site cGMP manufacturing
For supply chain continuity, we manufacture MCDB 131 at two separate facilities, located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP-manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
Specifications
Storage conditions: 2–8°C (protect from light) Shelf life: 12 months from date of manufacture |
|
1 X | |
Liquid | |
Sterile-filtered | |
No Glutamine, No HEPES | |
500 mL |
Human Microvascular Endothelial Cells (HMVEC), Human Omental Microvascular Cells, Hepatocytes, Myocytes, Smooth Muscle Cells | |
Mammalian Cell Culture | |
MCDB 131 Medium | |
Low Glucose, Phenol Red, Sodium Pyruvate | |
Gibco™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.