missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ VACUUBRAND™ MD1C Oil-Free Chemistry Diaphragm Vacuum Pump
An excellent choice for rotary evaporation and other laboratory applications
Supplier: BrandTech™ VACUUBRAND™ 20696600
Description
Suitable for benchtop rotary evaporators, and powerful enough for excellent results with high-boiling point solvents
- Extremely low maintenance, no oil changes, no oil mist
- Ultimate vacuum 1.5 Torr; free air capacity 25 liters per minute
- Corrosion resistance
- With an integrated gas ballast to provide consistent pump performance with condensed vapors
Specifications
0.08 hp | |
One phase motor, on/off switch, cable with plug, operating manual | |
CE | |
PTFE housing, ETFE pump head, PTFE Diaphragm, PTFE tubing, FFKM valve | |
CEE | |
15.2 lb. | |
104°F | |
10°C | |
50/60 Hz | |
17.5 cm | |
31.6 cm | |
IP42 |
0.8 A | |
1500 to 1800 | |
230 V 50/60 Hz | |
MD 1C | |
-10°C to +60°C | |
6.9 kg | |
40°C | |
23 L/min. | |
6.8 in. | |
12.4 in. | |
5.62 in. | |
230 V |