Mopec ME100 Dual Draft Countertop Grossing Station
This versatile, highly ventilated and compact  counter top workstation saves valuable floor space while ensuring a safe work environment. Mopec™ ME100 Dual Draft is designed for laboratories with both space constraints and the need for maximum ventilation.
Supplier:  Mopec ME100
Catalog No. 22444206
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Fixture with swing spout
  • Paper towel dispenser
  • One large and one small organizer bin
  • 30cm adjustable shelf
  • Energy saving, long life LED light
  • GFCI outlet
  • Inch/centimeter ruler
  • Magnetic tool bar
  • Perforated grid plates
  • Total draft chamber
Warranty
1 Year Warranty
Specifications
Specifications
Grossing Station
GFCI outlet
73 cm
110cm
30cm adjustable shelf
Total draft chamber
Fixed
121 cm