missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mopec ME300 Dual Draft Exhaust Grossing Station with Drain Chamber

This versatile ventilated workstation with leg frame offers more work space while ensuring a safe work environment. Mopec™ ME300 Dual Draft Exhaust Grossing Station with Drain Chamber is a medium sized grossing station designed for laboratories that need maximum ventilation of the work space.

Supplier:  Mopec ME300

Catalog No. ME300

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • Free standing unit
  • Down and back draft ventilation
  • Fixture with swing spout
  • Magnetic tool bar
  • Paper towel dispenser
  • One large and one small organizer bin
  • Inch and centimeter ruler
  • Recessed energy saving long life LED light
  • GFCI outlet
  • Perforated grid plates 125 in. holes
  • Back draft grill
  • Down draft chamber and leg frame

Warranty

1 Year Warranty

Specifications

Specifications

30 cm adjustable shelf
152cm
195cm
Down and back draft ventilation
Fixed
73 cm
Videos
SDS
Documents

Documents

Promotions

Promotions