missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mecillinam, 90+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J66774MC
Description
Effective against Gram-negative bacteria
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Mecillinam | |
Powder | |
156°C | |
100 mg | |
C15H23N3O3S | |
551°C | |
Soluble in water at approximately 1mg/mL | |
CC1(C(N2C(S1)C(C2=O)N=CN3CCCCCC3)C(=O)O)C | |
325.427 | |
CHEBI:51208 | |
95% |
Mecillinam | |
32887-01-7 | |
White | |
>110°C (230°F) | |
MFCD00056869 | |
14,389 | |
BWWVAEOLVKTZFQ-NTZNESFSSA-N | |
(2S,5R,6R)-6-(azepan-1-ylmethylideneamino)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid | |
36273 | |
325.43 |
Chemical Identifiers
32887-01-7 | |
325.427 | |
BWWVAEOLVKTZFQ-NTZNESFSSA-N | |
CHEBI:51208 | |
CC1(C(N2C(S1)C(C2=O)N=CN3CCCCCC3)C(=O)O)C |
C15H23N3O3S | |
MFCD00056869 | |
36273 | |
(2S,5R,6R)-6-(azepan-1-ylmethylideneamino)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
Safety and Handling
EINECSNumber : 251-277-1
RTECSNumber : XI0185000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only