missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Medegen Biohazardous Waste Bags, Autoclavable

These Biohazardous Waste Bags are autoclavable.

 View more versions of this product

Catalog No. 14-828-350


Explore available promotions

Description

Description

  • Autoclavable
Specifications

Specifications

Autoclavable Biohazardous Waste Bag
44 gal.
Meets 165 g Dart Impact Resistance Test
Indicator, flat pack, twist ties
0.045 mm
38 x 47 in. (96.52 x 119.38cm)
96.52 cm
119.38 cm
Autoclavable
166.6 L
Clear
Polypropylene
1.80 mil
38 in.
47 in.
SDS
Documents

Documents

Promotions

Promotions