Medegen Stainless-Steel Beakers
Will not rust, chip or peel
Supplier:  Medegen 80250
 View more versions of this product
Catalog No. 02-583B
Explore available promotions
Description
Description
  • Noncorrosive stainless steel
  • Can be heated
  • Seamless construction, smooth inner surface for easy cleaning
Specifications
Specifications
3 in.
250 mL
3.37 in.
Spout
Griffin
7.62 cm
Stainless Steel
8.55 cm
Beaker