Medegen Stainless-Steel Beakers
Will not rust, chip or peel
Supplier:  Medegen MDG84000
 View more versions of this product
Catalog No. 02-583G
Explore available promotions
Description
Description
  • Noncorrosive stainless steel
  • Can be heated
  • Seamless construction, smooth inner surface for easy cleaning
Specifications
Specifications
4.62 in.
4000 mL
8.12 in.
Spout
Griffin
11.73 cm
Stainless Steel
20.62 cm
Beaker