Learn More
Corning™ Medium 199 (Mod.)
Guaranteed low endotoxin levels ≤0.025 EU/mL
$207.43 - $207.43
Specifications
pH 7 to 7.4
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MT10060CV
View Documents
Corning™
10060CV
6 x 500 mL N/A
Only null left
Description
Description
  • Complete line of media and reagents for mammalian in vitro cell culture
  • Ready-to-use media is supplied in polyethylene teraphthalate (PET) plastic bottles
  • Sonically welded cap liners eliminate toxic resins or glues that could leach into media
  • Shipped at ambient temperature
Specifications
Specifications
7 to 7.4
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'