Gibco™ Human Large Vessel Endothelial Cell Basal Medium (formerly Medium 200) DFS Item
Medium 200
Supplier:  Gibco™ M200500
Catalog No. M200500
Explore available promotions
Description
Description
Human Large Vessel Endothelial Cell Basal Medium (formerly 'Medium 200') is a sterile, liquid tissue culture medium intended for use as one component in a complete culture environment for normal human large vessel endothelial cells. When supplemented with low serum growth supplement (LSGS), this medium supports the plating and proliferation of endothelial cells at culture densities between 2.5 x 103 cells/cm2 and 1 x 105 cells/cm2. Additional applications may include primary isolation of endothelial cells. This medium is also available without phenol red (Cat. No. M200PRF500).
Human Large Vessel Endothelial Cell Basal Medium is a basal culture medium containing essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. This medium does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. It is HEPES and bicarbonate buffered and designed for use in an incubator with an atmosphere of 5% CO2/95% air. To support the plating and proliferation of human large vessel endothelial cells, this medium must be supplemented with LSGS (Cat. No. S-003-10; kit = Cat. No. S-003-K).
Specifications
Specifications
Store in refrigerator (2–8°C) and protect from light.
Mammalian Cell Culture
Basal Medium
Sterile-filtered
Cascade Biologics™
Room Temperature
Endothelial Cells
Liquid
Sterile-filtered
HEPES, Phenol Red
500 mL
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.