missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Human Vascular Smooth Muscle Cell Basal Medium (formerly Medium 231)
Human Vascular Smooth Muscle Cell Basal Medium formerly Medium 231) is a sterile, liquid culture medium intended for use as one component in a complete culture environment for the growth of normal human vascular smooth muscle cells. When supplemented with SMGS, this medium will support the plating and proliferation of smooth muscle cells at densities between 2.5 x 103 cells/cm2 and 1 x 105 cells/cm2.
Supplier: Gibco™ M231500
Description
- Additional applications for use may include primary isolation of human smooth muscle cells
- Medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts; does not contain antibiotics, antimycotics, hormones, growth factors, or proteins
- HEPES and bicarbonate buffered; designed for use in an incubator with an atmosphere of 5% CO2/95% air
- To support the plating and proliferation of normal human vascular smooth muscle cells, Medium 231 must be supplemented with Smooth Muscle Growth Supplement (SMGS)
- To induce differentiation of normal human vascular smooth muscle cells, Human Vascular Smooth Muscle Cell Basal Medium must be supplemented with Smooth Muscle Differentiation Supplement (SMDS)
Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Mammalian Cell Culture | |
Basal Medium | |
Sterile-filtered | |
No Phenol Red | |
500 mL |
Smooth Muscle Cells | |
Liquid | |
Sterile-filtered | |
HEPES | |
Cascade Biologics™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.