Medix™ Thrombotic Pure Plus Test Kit DFS Item
Replacement for Unopette line for the counting of platelets
Supplier:  Medix™ BTP4015
Catalog No. 23230231
Explore available promotions
Description
Description
  • Microscopic counting of thrombocytes
  • Includes capillaries
Specifications
Specifications
Thrombotic Pure Plus Test Kit
Capillaries
Used for counting of Platelets, PLT (Unopette replacement), microscopic counting of thrombocytes (PLT)
1 Kit