missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Meglumine antimoniate

$230.03 - $230.03

Chemical Identifiers

CAS 133-51-7
Molecular Formula C7H17NO5·HO3Sb
Molecular Weight (g/mol) 365.97
InChI Key YPAUTKZODZLFQM-OVHUINQISA-K
PubChem CID 131664208
IUPAC Name antimony(3+);(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;trihydroxide;hydrate
SMILES CNCC(C(C(C(CO)O)O)O)O.O.[OH-].[OH-].[OH-].[Sb+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC461140250
View Documents
Thermo Scientific Chemicals
461140250
25 g
Each for $230.03
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

133-51-7
365.97
131664208
CNCC(C(C(C(CO)O)O)O)O.O.[OH-].[OH-].[OH-].[Sb+3]
C7H17NO5·HO3Sb
YPAUTKZODZLFQM-OVHUINQISA-K
antimony(3+);(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;trihydroxide;hydrate

Specifications

133-51-7
25 g
CNCC(C(C(C(CO)O)O)O)O.O.[OH-].[OH-].[OH-].[Sb+3]
365.97
365.97
C7H17NO5·HO3Sb
YPAUTKZODZLFQM-OVHUINQISA-K
antimony(3+);(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;trihydroxide;hydrate
131664208
Meglumine antimoniate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 205-108-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only