Integra™ Miltex™ MeisterHand™ Mayo Dissecting Scissors
distal cutting
Supplier: Integra™ Miltex™ MH5128
Description
- Stainless steel
- Non-glare matted finish
- Sharp/sharp scissors available in straight and curved patterns
- Tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
- SuperCut scissors have a razor sharp edge that cuts effortlessly through tissue and lower blade has micro fine serrations to help prevent tissue slippage; black ring handles
Order Info
Cold or chemical sterilization not recommended for tungsten carbide scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-sharp edge to nick or become irregular.
Specifications
Dissecting Scissors | |
9 in. | |
Stainless Steel |
Straight, Beveled | |
22.9 cm | |
Dissecting |