Integra™ Miltex™ MeisterHand™ Scalpel Handles
$45.18 - $179.29
Specifications
Blade Material Stainless Steel
Product Type Scalpel Handle
Description Non-glare Matte
View More Specs
Products 3
Catalog Number Mfr. No. Size For Use With (Equipment) Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Size For Use With (Equipment) Length (English) Length (Metric) Price Quantity  
12-460-633
View Documents
Integra™ Miltex™
MH410
# 3L # 10 to # 15C Surgical Blade 8.25 in. 21.2 cm
Each for $179.29
12-460-634
View Documents
Integra™ Miltex™
MH47
# 3 # 10 to # 15C Surgical Blade for Deep Surgery 5 in. 12.5 cm
Each for $45.18
12-460-635
View Documents
Integra™ Miltex™
MH48
# 4 # 20 to # 25 Blade for Deep Surgery 5.25 in. 13.5 cm
Each for $53.47
Description
Description
  • Extra fine
  • Stainless steel
  • With mm and cm graduations on No. 3 and No. 4
  • Non-glare mattefinish
Specifications
Specifications
Stainless Steel
Scalpel Handle
Non-glare Matte
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'