missing translation for 'onlineSavingsMsg'
Learn More

Melamine, 99%

CAS: 108-78-1 | C3H6N6 | 126.12 g/mol

$44.97 - $79.58

Chemical Identifiers

CAS 108-78-1
Molecular Formula C3H6N6
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006055
InChI Key JDSHMPZPIAZGSV-UHFFFAOYSA-N
Synonym melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr
PubChem CID 7955
ChEBI CHEBI:27915
IUPAC Name 1,3,5-triazine-2,4,6-triamine
SMILES NC1=NC(N)=NC(N)=N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125350050
SDS View Documents
Thermo Scientific Chemicals
125350050
5 g Glass bottle
Each for $44.97
Only null left
 
AC125351000
SDS View Documents
Thermo Scientific Chemicals
125351000
100 g Plastic bottle
Each for $56.42
Only null left
 
AC125350010
SDS View Documents
Thermo Scientific Chemicals
125350010
1 kg Plastic bottle
Each for $79.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-78-1
126.12
JDSHMPZPIAZGSV-UHFFFAOYSA-N
7955
1,3,5-triazine-2,4,6-triamine
C3H6N6
MFCD00006055
melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr
CHEBI:27915
NC1=NC(N)=NC(N)=N1

Specifications

108-78-1
White
Authentic
Glass bottle
MFCD00006055
26, 245
15, 5880
Solubility in water: 3g/L (20°C). Other solubilities: 0.6g/L in ethanol (20°C),sparingly soluble in glycol,glycerol,pyridine,insoluble in ether,benzene,carbon tetrachloride
NC1=NC(N)=NC(N)=N1
126.12
CHEBI:27915
99%
Melamine
354°C
1.5730g/mL
99%
C3H6N6
5 g
1.573
melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr
JDSHMPZPIAZGSV-UHFFFAOYSA-N
1,3,5-triazine-2,4,6-triamine
7955
126.12
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

EINECSNumber : 203-615-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only