BUCHI Melting and Boiling Point Instruments
Fast and accurate. BUCHI M-560/565 Melting Point Instruments have simple menus and programs that are easy to navigate on the intuitive LCD display, with either manual or automatic, real time video instrument.
Supplier: BUCHI 051998
Includes: Sample loader M-569, Calibration set M-565, melting point tubes (100 pieces), boiling point tubes (10 pieces), boiling capillary (10 pieces), sample holder, cleaning tool, CD demo melting point monitor, protection cover, USB cable (2m), operation manual
Description
- Fail-safe calibration with automated determination and storage of calibration results
- Automatic determination of melting and boiling points
- Highly accurate temperature regulation and measurement
- Optimized reproducibility with homogeneous packing of the capillary using the Sample Loader M-569
- Control of almost any type of sample by measuring melting point, boiling point or slip melting point.
- Double check for difficult samples by comparing manual and automated measurement
- Observation and replay of the phase transition using color display and video recording
- PC software included for easy management of methods and results
- Compliant with Pharmacopeia methods (European, USP, Japanese)
- Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories.
Compliance
Pharmacopeia methods (European, USP, Japanese); Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories.
Specifications
Automatic Melting Point Instrument | |
37 cm | |
100/240 V, 50/60 Hz, 150 W | |
7.8 in. | |
Melting Point instrument, Sample Loader M-569, Calibration Set M-565, melting point tubes (100), boiling point tubes (10), boiling capillary (10), sample holder, cleaning tool, CD demo melting point monitor, protective cover, USB cable (2m) | |
0.1°C (temp.) | |
4.5 kg | |
19 cm |
14.5 in. | |
TFT | |
Determining Melting Point, Boiling Point and Sling Melting Point | |
20 cm | |
M-565 | |
10°C to 400°C | |
7.4 in. |