missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bulwark™ Men's Excel FR™ ComforTouch™ Navy Premium Flame Resistant Coverall

Men's Excel FR™ ComforTouch™ Navy Premium Flame Resistant Coverall is a premium coverall that provides CAT 2 FR/AR protection and extra warmth for cold weather environments, without added bulk.

$613.09 - $613.09

Specifications

Product Type Flame-resistant Coveralls
Gender Men's
Material Cotton, Nylon
Color Navy
Disposable Reusable
View More Specs

Products 9
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-203-951
View Documents
Bulwark™
CLC8NVRGS
Small
Each for $613.09
 
17-203-945
View Documents
Bulwark™
CLC8NVRGM
Medium
Each for $613.09
 
17-204-026
View Documents
Bulwark™
CLC8NVRGL
Large
Each for $613.09
 
17-204-296
View Documents
Bulwark™
CLC8NVRGXL
X-Large
Each for $613.09
 
17-204-027
View Documents
Bulwark™
CLC8NVRGXXL
2X-Large
Each for $613.09
 
17-204-359
View Documents
Bulwark™
CLC8NVRG3XL
3X-Large
Each for $613.09
 
17-204-370
View Documents
Bulwark™
CLC8NVLNL
Large (Long)
Each for $613.09
 
17-204-154
View Documents
Bulwark™
CLC8NVLNXL
X-Large (Long)
Each for $613.09
 
17-204-161
View Documents
Bulwark™
CLC8NVLNXXL
2X-Large (Long)
Each for $613.09
 
Description

Description

  • This premium coverall provides CAT 3 FR/AR protection and extra warmth for cold weather environments, without added bulk.
  • It also accommodates snap on FR hoods for additional insulation from the elements.
  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 3 Protection
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Concealed Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Shell: Flame resistant, 7 oz. (235 g/m&superscript_2;) Twill, Water-repellent, Insulation Package: 14 oz.(474 g/m&superscript_2;)
  • Blend: Shell: 88% Cotton/ 12% Nylon, Lining: 100% Cotton, Insulation Package: 100% Modacrylic
  • Closure: Two-way, extra heavy-duty Nomex™ taped brass zipper front with concealed snap closures
  • Collar: Two-piece, insulated, stand-up collar with concealed snap closure
  • Pocket: Seven large patch pockets, two with hook-and-loop closure
  • Features: Full Nomex™ taped ankle-to-hip brass zippers, concealed snap closure
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 36
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Designed to be worn over other clothing. Fit over work boots.
  • Gender: Male
  • Materials: Cotton, Nylon
  • Brand: Bulwark™
Specifications

Specifications

Flame-resistant Coveralls
Cotton, Nylon
Reusable
Open
Collared
Snap
Non-sterile
Flame
36
7
Men's
Navy
Elastic (Stitched-in)
Open
Unhooded
Stitched
CAT 3
Two-Piece, Insulated, Stand-Up Collar
ASTM F1506, NFPA 2112, NFPA 70E
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'