missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bulwark™ Men's Midweight Excel FR™ ComforTouch™ Navy Premium Flame Resistant Coverall
Men's Midweight Excel FR™ ComforTouch™ Navy Premium Flame Resistant Coverall is a midweight premium coverall provides CAT 2 FR/AR protection with innovative features for extra comfort and convenience.
$221.96 - $221.96
Specifications
Product Type | Flame-resistant Coveralls |
---|---|
Gender | Men's |
Material | Cotton, Nylon |
Color | Navy |
Disposable | Reusable |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-204-184
|
Bulwark™
CLB6NVRGM |
Medium |
Each for $221.96
|
|
|||||
17-204-244
|
Bulwark™
CLB6NVRGL |
Large |
Each for $221.96
|
|
|||||
17-204-017
|
Bulwark™
CLB6NVRGXL |
X-Large |
Each for $221.96
|
|
|||||
17-204-099
|
Bulwark™
CLB6NVRGXXL |
2X-Large |
Each for $221.96
|
|
|||||
17-204-393
|
Bulwark™
CLB6NVRG3XL |
3X-Large |
Each for $221.96
|
|
|||||
17-204-183
|
Bulwark™
CLB6NVRG4XL |
4X-Large |
Each for $221.96
|
|
|||||
17-204-366
|
Bulwark™
CLB6NVLNL |
Large (Long) |
Each for $221.96
|
|
|||||
17-204-136
|
Bulwark™
CLB6NVLNXL |
X-Large (Long) |
Each for $221.96
|
|
|||||
17-204-139
|
Bulwark™
CLB6NVLNXXL |
2X-Large (Long) |
Each for $221.96
|
|
|||||
17-204-374
|
Bulwark™
CLB6NVLN3XL |
3X-Large (Long) |
Each for $221.96
|
|
|||||
17-204-124
|
Bulwark™
CLB6NVLN4XL |
4X-Large (Long) |
Each for $221.96
|
|
|||||
Description
- This midweight premium coverall provides CAT 2 FR/AR protection with innovative features for extra comfort and convenience.
- Eight total pockets keep your tools at hand, while elastic waist inserts and deep-pleated action back provide added mobility and comfort while you work.
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Concealed Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 9 oz. (305 g/m&superscript_2;) Twill
- Blend: 88% Cotton/ 12% Nylon
- Closure: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck
- Pocket: Two front swing pockets are topstitched and lined and two patch hip pockets have single concealed snap closure, One left sleeve pocket, two patch chest pockets have flaps and concealed snap closure and a rule pocket on right leg
- Features: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck, Two-piece, lay-flay collar, deep-pleated action back and elastic waist inserts, Hemmed sleeve with adjustable snap and side vent opening with concealed snap closure
- Product Collection: Excel FR Comfortouch
- Protection: ATPV Arc Rating: 12
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Designed to be worn over other clothing. Fit over work boots.
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
Flame-resistant Coveralls | |
Cotton, Nylon | |
Reusable | |
Open | |
Collared | |
2-way Front Zipper with Storm Flap | |
Non-sterile | |
Flame | |
NFPA 2112 |
Men's | |
Navy | |
Elastic (Stitched-in) | |
Open | |
Unhooded | |
Stitched | |
CAT 2 | |
12 | |
8 |