Learn More
Menthone, 98%, mixture of isomers, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125411000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Menthone | |
10458-14-7 | |
Colorless | |
207°C | |
Authentic | |
Glass bottle | |
1.4500 to 1.452 | |
100 g | |
14,201; 16,204 | |
10, 5663 | |
Solubility in water: slightly soluble. Other solubilities: soluble in organic solvents | |
CC1CCC(C(=O)C1)C(C)C | |
154.25 | |
CHEBI:36742 | |
98% |
90% | |
-6°C | |
0.8900g/mL | |
69°C | |
97.5% min. (GC) | |
C10H18O | |
MFCD00062998 | |
07, 38 | |
0.89 | |
isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone | |
NFLGAXVYCFJBMK-UHFFFAOYSA-N | |
5-methyl-2-propan-2-ylcyclohexan-1-one | |
6986 | |
154.25 | |
Liquid |
Chemical Identifiers
10458-14-7 | |
154.25 | |
NFLGAXVYCFJBMK-UHFFFAOYSA-N | |
6986 | |
5-methyl-2-propan-2-ylcyclohexan-1-one |
C10H18O | |
MFCD00062998 | |
isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone | |
CHEBI:36742 | |
CC1CCC(C(=O)C1)C(C)C |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 233-944-9
RUO – Research Use Only