missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Menthone, mixture of isomers, 98%, Thermo Scientific Chemicals
$111.77 - $759.68
Chemical Identifiers
CAS | 10458-14-7 |
---|---|
Molecular Formula | C10H18O |
Molecular Weight (g/mol) | 154.253 |
MDL Number | MFCD00062998 |
InChI Key | NFLGAXVYCFJBMK-UHFFFAOYSA-N |
Synonym | isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone |
PubChem CID | 6986 |
ChEBI | CHEBI:36742 |
IUPAC Name | 5-methyl-2-propan-2-ylcyclohexan-1-one |
SMILES | CC1CCC(C(=O)C1)C(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1766518
|
Thermo Scientific Chemicals
A1766518 |
50 g |
Each for $111.77
|
|
||||||
AAA1766530
|
Thermo Scientific Chemicals
A1766530 |
250 g |
Each for $279.41
|
|
||||||
AAA176650B
|
Thermo Scientific Chemicals
A176650B |
1000 g |
Each for $759.68
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10458-14-7 | |
154.253 | |
NFLGAXVYCFJBMK-UHFFFAOYSA-N | |
6986 | |
5-methyl-2-propan-2-ylcyclohexan-1-one |
C10H18O | |
MFCD00062998 | |
isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone | |
CHEBI:36742 | |
CC1CCC(C(=O)C1)C(C)C |
Specifications
10458-14-7 | |
∼207°C | |
C10H18O | |
50 g | |
NFLGAXVYCFJBMK-UHFFFAOYSA-N | |
5-methyl-2-propan-2-ylcyclohexan-1-one | |
6986 | |
154.25 | |
Menthone, mixture of isomers |
0.894 | |
72°C (162°F) | |
MFCD00062998 | |
isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone | |
CC1CCC(C(=O)C1)C(C)C | |
154.253 | |
CHEBI:36742 | |
98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 233-944-9
RTECSNumber : OS9541500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only