missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mercuric Nitrate, 0.282 N, 1 mL = 10 mg Cl-, Ricca Chemical
Supplier: Ricca Chemical Company 47501
Specifications
Mercuric Nitrate | |
3.32,93.0,1.71 | |
H2HgN2O7 | |
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
342.61 | |
0.282N | |
4 L |
7783-34-8,7732-18-5,7697-37-2 | |
3.45,97.0,1.77 | |
MFCD00149736 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
mercury(2+) hydrate dinitrate | |
3084029 | |
Amber Glass Bottle |