missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mercury Metal, Instrument Grade, Reagents
Mercury Metal, Instrument Grade, Instrument, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C224400025A5
Specifications
Silver-White | |
-38.87°C | |
356.6°C | |
Hg | |
2.5 kg | |
QSHDDOUJBYECFT-UHFFFAOYSA-N | |
mercury | |
Poly Bottle |
Mercury Metal, Instrument Grade | |
7439-97-6 | |
200.59 | |
MFCD00011035 | |
Hg | |
[Hg] | |
200.59 | |
Liquid |
Chemical Identifiers
7439-97-6 | |
200.59 | |
QSHDDOUJBYECFT-UHFFFAOYSA-N | |
mercury |
Hg | |
MFCD00011035 | |
Hg | |
[Hg] |