Restek™ Merlin Microseal Replacement Septa
Withstands thousands of injections
Supplier: Restek™ RK22264
Description
- Eliminates septum coring and crumbling
- Reliable performance during extended runs
- O-ring detail seals needle during injection
- Duckbill seals more tightly at higher pressures
- Low insertion force for easy manual injections
- Reduced septum bleed and ghost peaks
Note:
- Merlin microseal septa requires appropriate adaptor kit or nut for specific GC
- Choose standard microseal kits/nuts for use with needles and traditional SPME fibers
- Choose SPME Arrow microseal kits/nuts for use with SPME Arrow
Specifications
26 Gauge or 23/26 Gauge Tapered Needles (5 to 100 Psi) | |
Replacement Septum |