Learn More
Mesityl oxide, 90+%, remainder 4-methyl-4-penten-2-one
CAS: 141-79-7 | C6H10O | 98.145 g/mol
$102.03 - $617.86
Chemical Identifiers
| CAS | 141-79-7 |
|---|---|
| Molecular Formula | C6H10O |
| Molecular Weight (g/mol) | 98.145 |
| MDL Number | MFCD00008900 |
| InChI Key | SHOJXDKTYKFBRD-UHFFFAOYSA-N |
| Synonym | mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone |
| PubChem CID | 8858 |
| IUPAC Name | 4-methylpent-3-en-2-one |
| SMILES | CC(=CC(=O)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA15111AE
|
Thermo Scientific Chemicals
A15111AE |
100 mL |
N/A
|
|
|||||
|
AAA15111AK
|
Thermo Scientific Chemicals
A15111AK |
250 mL |
N/A
|
|
|||||
|
AAA15111AP
|
Thermo Scientific Chemicals
A15111AP |
500 mL |
N/A
|
|
|||||
|
AAA15111AU
|
Thermo Scientific Chemicals
A15111AU |
1000 mL |
N/A
|
|
|||||
Description
Mesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.
Solubility
Soluble in water(30g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
| 141-79-7 | |
| 98.145 | |
| SHOJXDKTYKFBRD-UHFFFAOYSA-N | |
| 8858 | |
| CC(=CC(=O)C)C |
| C6H10O | |
| MFCD00008900 | |
| mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone | |
| 4-methylpent-3-en-2-one |
Specifications
| 141-79-7 | |
| 0.856 | |
| 30°C (86°F) | |
| honigähnlich | |
| 1.444 | |
| 100 mL | |
| 1361550 | |
| mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone | |
| SHOJXDKTYKFBRD-UHFFFAOYSA-N | |
| 4-methylpent-3-en-2-one | |
| 8858 | |
| ≥90% | |
| Mesityl oxide |
| -59°C | |
| 129°C to 130°C | |
| ≥90% | |
| C6H10O | |
| MFCD00008900 | |
| UN1229 | |
| 14,5908 | |
| Soluble in water(30g/L). | |
| CC(=CC(=O)C)C | |
| 98.145 | |
| 98.15 | |
| Liquid | |
| remainder 4-methyl-4-penten-2-one |
Safety and Handling
GHS H Statement
H226-H302-H312-H332
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
H226-H302+H312+H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MESITYL OXIDE
EINECSNumber : 205-502-5
RTECSNumber : SB4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only