missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 608-36-6 |
---|---|
Molecular Formula | C4H2Br2O4 |
Molecular Weight (g/mol) | 273.87 |
MDL Number | MFCD00066439 |
InChI Key | FJWGRXKOBIVTFA-XIXRPRMCSA-L |
Synonym | 2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi |
PubChem CID | 641611 |
IUPAC Name | (2S,3R)-2,3-dibromobutanedioic acid |
SMILES | [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC147575000
|
Thermo Scientific Chemicals
147575000 |
500g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
608-36-6 | |
273.87 | |
FJWGRXKOBIVTFA-XIXRPRMCSA-L | |
641611 | |
[O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O |
C4H2Br2O4 | |
MFCD00066439 | |
2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi | |
(2S,3R)-2,3-dibromobutanedioic acid |
Specifications
608-36-6 | |
98% | |
C4H2Br2O4 | |
MFCD00066439 | |
02,625 | |
2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi | |
FJWGRXKOBIVTFA-XIXRPRMCSA-L | |
(2S,3R)-2,3-dibromobutanedioic acid | |
641611 | |
98% |
Authentic | |
Plastic bottle | |
HO2CCH(Br)CH(Br)CO2H | |
500g | |
14,3029 | |
Solubility in water: 20g/L (17°C). Other solubilities: soluble in alcohol and ether, sparingly soluble in chloroform | |
[O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O | |
273.87 | |
275.88 | |
meso-2,3-Dibromosuccinic acid |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only