missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-2,3-Dimercaptosuccinic acid, 98%
CAS: 304-55-2 | C4H6O4S2 | 182.21 g/mol
$161.65 - $527.88
Chemical Identifiers
| CAS | 304-55-2 |
|---|---|
| Molecular Formula | C4H6O4S2 |
| Molecular Weight (g/mol) | 182.21 |
| MDL Number | MFCD00064799 |
| InChI Key | ACTRVOBWPAIOHC-XIXRPRMCSA-N |
| Synonym | succimer, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, dmsa, dim-sa, 2r,3s-rel-2,3-dimercaptosuccinic acid, succimero, succimerum, chemet, dimercaptosuccinic acid |
| PubChem CID | 2724354 |
| ChEBI | CHEBI:63623 |
| IUPAC Name | (2S,3R)-2,3-bis(sulfanyl)butanedioic acid |
| SMILES | C(C(C(=O)O)S)(C(=O)O)S |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC226670050
|
Thermo Scientific Chemicals
226670050 |
5 g | Glass bottle |
Each for $161.65
|
|
||||
|
AC226670250
|
Thermo Scientific Chemicals
226670250 |
25 g | Glass bottle |
Each for $527.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 304-55-2 | |
| 182.21 | |
| ACTRVOBWPAIOHC-XIXRPRMCSA-N | |
| 2724354 | |
| (2S,3R)-2,3-bis(sulfanyl)butanedioic acid |
| C4H6O4S2 | |
| MFCD00064799 | |
| succimer, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, dmsa, dim-sa, 2r,3s-rel-2,3-dimercaptosuccinic acid, succimero, succimerum, chemet, dimercaptosuccinic acid | |
| CHEBI:63623 | |
| C(C(C(=O)O)S)(C(=O)O)S |
Specifications
| 304-55-2 | |
| White | |
| Authentic | |
| Glass bottle | |
| HO2CCH(SH)CH(SH)CO2H | |
| 5 g | |
| succimer, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, dmsa, dim-sa, 2r,3s-rel-2,3-dimercaptosuccinic acid, succimero, succimerum, chemet, dimercaptosuccinic acid | |
| C(C(C(=O)O)S)(C(=O)O)S | |
| 182.21 | |
| CHEBI:63623 | |
| 98% | |
| meso-2, 3-Dimercaptosuccinic acid, 98% |
| 196°C to 198°C | |
| 1% max. (1 g, 105°C, 3 hrs) | |
| 98% | |
| C4H6O4S2 | |
| MFCD00064799 | |
| 15,8995 | |
| ACTRVOBWPAIOHC-XIXRPRMCSA-N | |
| (2S,3R)-2,3-bis(sulfanyl)butanedioic acid | |
| 2724354 | |
| 182.21 | |
| Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child.
GHS Signal Word: Warning
EINECSNumber : 206-155-2
RTECSNumber : WM7650000
TSCA : TSCA
RUO – Research Use Only