missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
meso-Erythritol, 99%, Thermo Scientific Chemicals
$169.79 - $480.29
Chemical Identifiers
CAS | 149-32-6 |
---|---|
Molecular Formula | C4H10O4 |
Molecular Weight (g/mol) | 122.12 |
MDL Number | MFCD00004710 |
InChI Key | UNXHWFMMPAWVPI-UHFFFAOYNA-N |
Synonym | erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite |
PubChem CID | 222285 |
ChEBI | CHEBI:17113 |
SMILES | OCC(O)C(O)CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC117820250
|
Thermo Scientific Chemicals
117820250 |
25 g | Plastic Bottle |
Each for $169.79
|
|
||||
AC117821000
|
Thermo Scientific Chemicals
117821000 |
100 g | Plastic bottle |
Each for $480.29
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
149-32-6 | |
122.12 | |
UNXHWFMMPAWVPI-UHFFFAOYNA-N | |
222285 | |
OCC(O)C(O)CO |
C4H10O4 | |
MFCD00004710 | |
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite | |
CHEBI:17113 |
Specifications
149-32-6 | |
White | |
Authentic | |
Plastic Bottle | |
HOCH2[CH(OH)]2CH2OH | |
25 g | |
15,3733 | |
(10% in water) Clear colorless | |
OCC(O)C(O)CO | |
122.12 | |
CHEBI:17113 | |
99% | |
meso-Erythritol, 99% |
117.0°C to 121.0°C | |
329.0°C to 331.0°C | |
98.5% min. (ELSD) (HPLC) | |
C4H10O4 | |
MFCD00004710 | |
01,525 | |
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite | |
UNXHWFMMPAWVPI-UHFFFAOYNA-N | |
(2S,3R)-butane-1,2,3,4-tetrol | |
222285 | |
122.12 | |
Crystalline Powder or Crystals |
Safety and Handling
EINECSNumber : 205-737-3
RTECSNumber : KF2000000
RUO – Research Use Only