MilliporeSigma™ Supelco™ Metal Packed GC Column (fits Agilent, Configuration A), Phase: 0.2 Carbowax 1500, Matrix 80/100 Carbopack C Support
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.
Supplier: MilliporeSigma™ Supelco™ 12506U
Specifications
Packed GC | |
Gas Chromatography (GC) | |
Stainless Steel Column | |
2.1 mm |
0.125 in. | |
1.8 m | |
Carbowax 1500, Matrix 80/100 Carbopack C Support |