MilliporeSigma™ Supelco™ Metal Packed GC Column (fits Agilent™, Configuration A), Phase: 25 SP™-2100, Matrix 80/100 Chromosorb™ P AW Support
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.
Supplier: MilliporeSigma™ Supelco™ 12995U
Specifications
Packed GC | |
Gas Chromatography (GC) | |
Stainless Steel Column | |
0.75 mm |
0.0625 in. | |
1.7 m | |
SP-2100, Matrix 80/100 Chromosorb P AW Support |
SP is a registered trademark of Sigma-Aldrich Co. LLC