missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MetFormin hydrochloride, 98%, For HPLC analysis, MP Biomedicals™
$66.85 - $300.83
Chemical Identifiers
CAS | 1115-70-4 |
---|---|
Molecular Formula | C4H12ClN5 |
Molecular Weight (g/mol) | 165.63 |
MDL Number | MFCD00012582 |
InChI Key | OETHQSJEHLVLGH-UHFFFAOYSA-N |
Synonym | metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform |
PubChem CID | 14219 |
IUPAC Name | hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride |
SMILES | [H+].[Cl-].CN(C)C(=N)N=C(N)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN15169101
|
MP Biomedicals Inc
0215169101 |
1 g |
Each for $66.85
|
|
|||||
ICN15169105
|
MP Biomedicals Inc
0215169105 |
5 g |
Each for $157.69
|
|
|||||
ICN15169110
|
MP Biomedicals Inc
0215169110 |
10 g |
Each for $300.83
|
|
|||||
Chemical Identifiers
1115-70-4 | |
165.63 | |
OETHQSJEHLVLGH-UHFFFAOYSA-N | |
14219 | |
[H+].[Cl-].CN(C)C(=N)N=C(N)N |
C4H12ClN5 | |
MFCD00012582 | |
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform | |
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride |
Specifications
1115-70-4 | |
C4H12ClN5 | |
1 g | |
OETHQSJEHLVLGH-UHFFFAOYSA-N | |
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride | |
14219 | |
0.98 | |
Metformin hydrochloride, For HPLC analysis |
White | |
MFCD00012582 | |
metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform | |
[H+].[Cl-].CN(C)C(=N)N=C(N)N | |
165.63 | |
165.6 | |
Crystalline Powder |