missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methacrolein (stabilized with HQ) 90.0+%, TCI America™
Supplier: TCI America M007825ML
Specifications
Methacrolein (stabilized with HQ) | |
-81°C | |
68°C | |
MFCD00006974 | |
2396 | |
STNJBCKSHOAVAJ-UHFFFAOYSA-N | |
2-methylprop-2-enal | |
6562 | |
≥90.0% (GC) |
78-85-3 | |
Yellow | |
C4H6O | |
25 mL | |
methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde | |
CC(=C)C=O | |
70.09 | |
70.09 | |
Liquid |
Chemical Identifiers
78-85-3 | |
70.09 | |
STNJBCKSHOAVAJ-UHFFFAOYSA-N | |
6562 | |
CC(=C)C=O |
C4H6O | |
MFCD00006974 | |
methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde | |
2-methylprop-2-enal |
Safety and Handling
EINECSNumber : (2)-0522
RTECSNumber : OZ2625000
TSCA : Yes
Recommended Storage : Refrigerator