missing translation for 'onlineSavingsMsg'
Learn More

Methacrylamide, 98%

CAS: 79-39-0 | C4H7NO | 85.106 g/mol

$54.37 - $100.79

Chemical Identifiers

CAS 79-39-0
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00008018
InChI Key FQPSGWSUVKBHSU-UHFFFAOYSA-N
Synonym methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
PubChem CID 6595
ChEBI CHEBI:51759
IUPAC Name 2-methylprop-2-enamide
SMILES CC(=C)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1501322
View Documents
Thermo Scientific Chemicals
L1501322
100 g
Each for $54.37
Only null left
 
AAL1501336
View Documents
Thermo Scientific Chemicals
L1501336
500 g
Each for $100.79
Only null left
 
Description

Description

Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

Solubility
Soluble in water (202g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
Specifications

Chemical Identifiers

79-39-0
85.106
FQPSGWSUVKBHSU-UHFFFAOYSA-N
6595
2-methylprop-2-enamide
C4H7NO
MFCD00008018
methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
CHEBI:51759
CC(=C)C(=O)N

Specifications

79-39-0
1.115
C4H7NO
100 g
methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
FQPSGWSUVKBHSU-UHFFFAOYSA-N
2-methylprop-2-enamide
6595
85.11
Methacrylamide
107°C to 112°C
215°C
MFCD00008018
605397
Soluble in water (202g/L at 20°C).
CC(=C)C(=O)N
85.106
CHEBI:51759
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P309+P311-P312-P330-P501c

H302-H319-H335-H371-H373

EINECSNumber : 201-202-3

RTECSNumber : UC6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only