missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methacryloyl chloride, may contain up to ca. 15% cyclic dimer, 97%, stab., Thermo Scientific Chemicals

$1798.86 - $1798.86

Chemical Identifiers

CAS 920-46-7
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.533
MDL Number MFCD00000716
InChI Key VHRYZQNGTZXDNX-UHFFFAOYSA-N
Synonym methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
PubChem CID 13528
IUPAC Name 2-methylprop-2-enoyl chloride
SMILES CC(=C)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1451136
View Documents
Thermo Scientific Chemicals
L1451136
500 g
Each for $1,798.86
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacryloyl chloride is used in the preparation of polymeric materials. It reacts with L-histidine and forms functional monomer. It is also used to prepare monomer 2-methacrylamido-caprolactam, amide monomers by amidation.

Solubility
Miscible with alcohols, ethers and organic solvents. Slightly miscible with water.

Notes
Incompatible with oxidizing agents and strong bases.
Specifications

Chemical Identifiers

920-46-7
104.533
VHRYZQNGTZXDNX-UHFFFAOYSA-N
13528
CC(=C)C(=O)Cl
C4H5ClO
MFCD00000716
methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
2-methylprop-2-enoyl chloride

Specifications

920-46-7
1.07
2°C (35°F)
C4H5ClO
MFCD00000716
UN3488
Moisture sensitive
Miscible with alcohols,ethers and organic solvents. Slightly miscible with water.
CC(=C)C(=O)Cl
104.533
104.54
Methacryloyl chloride, stabilized
-60°C
95°C to 96°C
Acrid
1.444
500 g
878175
methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
VHRYZQNGTZXDNX-UHFFFAOYSA-N
2-methylprop-2-enoyl chloride
13528
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H330-H314-H318-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 213-058-9

RTECSNumber : OZ5791000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only