Learn More
Methacryloyl chloride, may contain up to ca. 15% cyclic dimer, 97%, stab., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1451136
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethacryloyl chloride is used in the preparation of polymeric materials. It reacts with L-histidine and forms functional monomer. It is also used to prepare monomer 2-methacrylamido-caprolactam, amide monomers by amidation.
Solubility
Miscible with alcohols, ethers and organic solvents. Slightly miscible with water.
Notes
Incompatible with oxidizing agents and strong bases.
Specifications
Methacryloyl chloride | |
920-46-7 | |
1.07 | |
2°C (35°F) | |
C4H5ClO | |
MFCD00000716 | |
UN3488 | |
Moisture sensitive | |
Miscible with alcohols,ethers and organic solvents. Slightly miscible with water. | |
CC(=C)C(=O)Cl | |
104.533 | |
104.54 |
stabilized | |
-60°C | |
95°C to 96°C | |
Acrid | |
1.444 | |
500 g | |
878175 | |
methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride | |
VHRYZQNGTZXDNX-UHFFFAOYSA-N | |
2-methylprop-2-enoyl chloride | |
13528 | |
97% |
Chemical Identifiers
920-46-7 | |
104.533 | |
VHRYZQNGTZXDNX-UHFFFAOYSA-N | |
13528 | |
CC(=C)C(=O)Cl |
C4H5ClO | |
MFCD00000716 | |
methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride | |
2-methylprop-2-enoyl chloride |
Safety and Handling
GHS H Statement
H225-H330-H314-H318-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 213-058-9
RTECSNumber : OZ5791000
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only