missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 125610010

 View more versions of this product

Catalog No. AC125610010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methanesulfonic acid
19.0°C
167.0°C (10.0 mmHg)
Authentic
Glass bottle
1.4252 to 1.4315
MFCD00007518
04, 4
1.481
methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methanesulfonic acid
6395
CHEBI:27376
99%
Liquid
75-75-2
1.4810g/mL
189°C
99%
CH4O3S
CH3SO3H
1 L
01,666; 02,270; 04,326; 10,256; 11,321; 12,212; 13,176
15, 6026
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and ethanol, insoluble in hexane and methylcyclopentane, 1.50wt% benzene -0.23wt% 0-chlorotoluene (26-28°C), 0.38wt% toluene -0.47wt% ethyl disulfide (26-28°C)
CS(=O)(=O)O
96.1
11.6 mPa.s (25°C)
96.1
Extra Pure

Chemical Identifiers

75-75-2
96.1
AFVFQIVMOAPDHO-UHFFFAOYSA-N
6395
methanesulfonic acid
CH4O3S
MFCD00007518
methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
CHEBI:27376
CS(=O)(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
D

GHS Signal Word: Danger

EINECSNumber : 200-898-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only