Learn More
Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125641000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methanesulfonyl chloride | |
-33.0°C | |
161.0°C | |
Authentic | |
Glass bottle | |
1.4435 to 1.4455 | |
MFCD00007454 | |
04,5 | |
1.48 | |
mesyl chloride, methylsulfonyl chloride, methyl sulfochloride, methanesulphonyl chloride, chloro methyl sulfone, methanesulfuryl chloride, methanesulfonic acid chloride, methyl sulfonyl chloride, chloromethyl sulfone, mesylchloride | |
QARBMVPHQWIHKH-UHFFFAOYSA-N | |
methanesulfonyl chloride | |
31297 | |
114.55 | |
Liquid |
124-63-0 | |
1.4800g/mL | |
110°C | |
99.4% min. (GC) | |
CH3ClO2S | |
CH3SO2Cl | |
100 mL | |
01,662; 02,268; 04,326; 05,435; 06,362; 07,225; 11,322; 13,176; 17,180 | |
15,6027 | |
Solubility in water: insoluble (reacts with water). Other solubilities: soluble in ethanol, ether, acetone and other, common organic solvents | |
CS(Cl)(=O)=O | |
114.54 | |
1.97 mPa.s (25°C) | |
99.5% |
Chemical Identifiers
124-63-0 | |
114.54 | |
QARBMVPHQWIHKH-UHFFFAOYSA-N | |
31297 | |
CS(Cl)(=O)=O |
CH3ClO2S | |
MFCD00007454 | |
mesyl chloride, methylsulfonyl chloride, methyl sulfochloride, methanesulphonyl chloride, chloro methyl sulfone, methanesulfuryl chloride, methanesulfonic acid chloride, methyl sulfonyl chloride, chloromethyl sulfone, mesylchloride | |
methanesulfonyl chloride |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
May cause respiratory irritation.
May be corrosive
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Danger
EINECSNumber : 204-706-1
RUO – Research Use Only