missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, Absolute, Reagent, ACS, 99.8%, Spectrum™ Chemical

M1240, 67-56-1, CH3OH

Supplier:  Spectrum Chemical Mfg Cor M12404LTGLCS4

 View more versions of this product

Catalog No. 18604358


Explore available promotions

Description

Description

Spectrum™ Chemical Methanol, Absolute, Reagent, ACS , is also called methyl alcohol or ′wood alcohol′ it was one of the by products of distillation of wood. As the simplest alcohol around, it is light, flammable, colorless liquid with a strong and distinctive odor. It has many uses including acting as an antifreeze, (due to it′s polar liquid properties), solvent, fuel as well as a denaturant for ethanol. As an ACS grade Reagent, Spectrum Chemical manufactured methanol is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications

Specifications

67-56-1
4 x 4 L
0.001%
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
99.8%
100%
Amber Glass Bottle
CH4O
UN1230
CO
32.04

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
CO
CH4O
MFCD00004595
methanol
Videos
SDS
Documents

Documents

Promotions

Promotions