missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, ACS Reagent Grade, Absolute, Ricca Chemical

$134.88 - $1528.53

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
482932
SDS View Documents
Ricca Chemical Company
482932
1 L Natural Poly Bottle
Each for $134.88
 
48291
View Documents
Ricca Chemical Company
48291
4 L Natural Poly Bottle
Each for $366.09
 
48295HP
View Documents
Ricca Chemical Company
48295HP
20 L Hard Pack
Each for $1,528.53
 
Description

Description

Specifications

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
CHEBI:17790
CO
CH4O
MFCD00004595
887
methanol

Specifications

67-56-1
98.0
1 L
Liquid
max. 0.0003 meq/g
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:17790
ACS
Colorless
100.0
Natural Poly Bottle
max. 0.001%
CH4O
max. 0.1%
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
≥99.8%
Methanol, Absolute
Videos
Safety and Handling

Safety and Handling

ShelfLife : 36 months

SDS
missing translation for 'documents'

missing translation for 'documents'