Learn More
Methanol (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
$242.69 - $5795.33
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A452-1
|
Thermo Fisher Scientific
BPA4521 |
1 L | Wide Neck Amber Bottle |
Each for $242.69
Case of 6 Each for $1,456.14
|
|
||||
A452-4
|
Thermo Fisher Scientific
BPA4524 |
4 L | Glass Bottle |
Each for $319.08
Case of 4 Each for $1,276.32
|
|
||||
A452SK-4
|
Thermo Fisher Scientific
FLA452SK4 |
4 L | Safe-Cote™ Amber Glass |
Each for $416.50
Case of 4 Each for $1,666.00
|
|
||||
A452N119
|
Fisher Chemical
A452N119 |
19 L | NOWPak™ I |
Each for $5,795.33
|
|
||||
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-98°C | |
0.025 AU max. at 254nm, 0.15 AU max. at 230nm, 0.30 AU max. at 220nm, 1.00 AU max. at 205nm | |
64.7°C | |
Pass Test | |
Liquid | |
Assay: 99.9% min by GC | |
0.0003mEq/g max. | |
0.0002mEq/g max. | |
MFCD00004595 | |
0.55 cP at 20°C | |
0.1% max. | |
Solubility in water: miscible | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.9% | |
Methanol |
67-56-1 | |
Colorless | |
1 L | |
Wide Neck Amber Bottle | |
3ppm max. | |
0.791 | |
CH4O | |
CH3OH | |
128 hPa at 20°C | |
1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
Mobile phase for HPLC applications |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and eyes. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid